Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyldiphenylamine, 99%

$42.64 - $140.14

Chemical Identifiers

Chemical Name or Material 3-Methyldiphenylamine
CAS 1205-64-7
Molecular Formula C13H13N
Formula Weight 183.25
MDL Number MFCD00008530
Synonym N-Phenyl-m-toluidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2544109

  SDS
alfa aesar™
B2544109
10g Each for $42.64

AAB2544118

  SDS
alfa aesar™
B2544118
50g Each for $140.14
Description & Specifications

Specifications

Formula Weight 183.25
Assay 99%
Molecular Formula C13H13N
CAS 1205-64-7
Flash Point 147°C (297°F)
Boiling Point 317°C
Chemical Name or Material 3-Methyldiphenylamine
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 25° to 27°C
TSCA TSCA
Health Hazard 2 GHS H Statement
H302-H312-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Quantity 10g
Health Hazard 1 Warning
MDL Number MFCD00008530
Synonym N-Phenyl-m-toluidine
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H312-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.