Alfa Aesar™ 3-Methylcyclopentane-1,2-dione, 98+%
Manufacturer:  Alfa Aesar™ A1025509
 View more versions of this product
Catalog No. AAA1025509
Specifications
Specifications
3-Methylcyclopentane-1,2-dione
10g
C6H8O2
2076520
OACYKCIZDVVNJL-UHFFFAOYSA-N
3-methylcyclopentane-1,2-dione
61209
≥98%
103°C to 107°C
765-70-8
MFCD00001417
3-methyl-1,2-cyclopentanedione, 1,2-cyclopentanedione, 3-methyl, methylcyclopentenolone diketo form, fema no. 2700, methylcyclopentenolone natural, kentonarome, benzil-related compound, 45, 3-methyl-1,2-cyclopentanedione maple lactone, maple lactone mcp, methyl-cyclopentenolone
CC1CCC(=O)C1=O
112.128
112.13
Chemical Identifiers
765-70-8
112.128
OACYKCIZDVVNJL-UHFFFAOYSA-N
61209
CC1CCC(=O)C1=O
C6H8O2
MFCD00001417
3-methyl-1,2-cyclopentanedione, 1,2-cyclopentanedione, 3-methyl, methylcyclopentenolone diketo form, fema no. 2700, methylcyclopentenolone natural, kentonarome, benzil-related compound, 45, 3-methyl-1,2-cyclopentanedione maple lactone, maple lactone mcp, methyl-cyclopentenolone
3-methylcyclopentane-1,2-dione
Safety and Handling
Safety and Handling
TSCA : TSCA
SDS Documents