missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methylcyclohexanol (cis- and trans- mixture), TCI America™

Manufacturer:  TCI America M0195500ML

 View more versions of this product

Catalog No. M0195500ML



Specifications

Specifications

3-Methylcyclohexanol (cis- and trans- mixture)
500mL
C7H14O
3-methylcyclohexanol, m-methylcyclohexanol, cyclohexanol, 3-methyl, hexahydro-m-cresol, cyclohexanol, m-methyl, 3-methylcyclohexanol, mixed isomers, 3-methyl-cyclohexanol, cis-3-methylcyclohexanol, trans-3-methylcyclohexanol, 3-methylcyclohexanol, mixture of cis and trans
CC1CCCC(C1)O
114.188
114.19
≥98.0% (GC)
Colorless
591-23-1
MFCD00001446
HTSABYAWKQAHBT-UHFFFAOYSA-N
3-methylcyclohexan-1-ol
11566
Clear Liquid at 20°C

Chemical Identifiers

591-23-1
114.188
HTSABYAWKQAHBT-UHFFFAOYSA-N
11566
CC1CCCC(C1)O
C7H14O
MFCD00001446
3-methylcyclohexanol, m-methylcyclohexanol, cyclohexanol, 3-methyl, hexahydro-m-cresol, cyclohexanol, m-methyl, 3-methylcyclohexanol, mixed isomers, 3-methyl-cyclohexanol, cis-3-methylcyclohexanol, trans-3-methylcyclohexanol, 3-methylcyclohexanol, mixture of cis and trans
3-methylcyclohexan-1-ol
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2301

RTECSNumber : GW0200000

TSCA : Yes

Documents