Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methylcinnamic acid, predominantly trans, 98%

$248.62 - $248.62

Chemical Identifiers

Chemical Name or Material 3-Methylcinnamic acid
CAS 3029-79-6
Name Note predominantly trans
Molecular Formula C10H10O2
Formula Weight 162.19
MDL Number MFCD00016844
Beilstein 2042550
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2091409

  SDS
alfa aesar™
B2091409
10g Each for $248.62
Description & Specifications

Specifications

Formula Weight 162.19
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 10g
MDL Number MFCD00016844
Assay 98%
CAS 3029-79-6
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Beilstein 2042550
Chemical Name or Material 3-Methylcinnamic acid
Melting Point 115° to 119°C
Molecular Formula C10H10O2
Name Note predominantly trans
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.