missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Riedel-de Haen™ 3-Methylbutanol, Puriss. p.a., ACS Reagent, ≥98.5% (GC), Honeywell Riedel-de Haën™

Puriss. p.a., ACS Reagent,≥98.5% (GC)

$225.14 - $225.14

Chemical Identifiers

CAS 123-51-3
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00002934
InChI Key PHTQWCKDNZKARW-UHFFFAOYSA-N
Synonym 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
PubChem CID 31260
ChEBI CHEBI:15837
IUPAC Name 3-methylbutan-1-ol
SMILES CC(C)CCO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60047132 View Documents Honeywell Riedel-de Haen™
590901L
1L Glass bottle Each for $225.14
 
Description

Description

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
Specifications

Chemical Identifiers

123-51-3
88.15
PHTQWCKDNZKARW-UHFFFAOYSA-N
31260
3-methylbutan-1-ol
C5H12O
MFCD00002934
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
CHEBI:15837
CC(C)CCO

Specifications

Liquid
Glass bottle
Colorless
1L
C5H12O
MFCD00002934
1718835
PHTQWCKDNZKARW-UHFFFAOYSA-N
3-methylbutan-1-ol
31260
88.15g/mol
3-Methylbutanol
0.809g/mL at 20°C
131°C to 132°C
-117°C
123-51-3
(CH3)2CHCH2CH2OH
UN1105
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
CC(C)CCO
88.15
CHEBI:15837
≥98.5% (GC)
Safety and Handling

Safety and Handling

P210-P261-P280-P304 + P340 + P312-P337 + P313-P403 + P235

DOTInformation : class 3 - PG 3 - Pentanols

RTECSNumber : 204-633-5