Please login to your online account to display your discounted pricing

3-Methylbut-2-enyl formate, Maybridge

Chemical Identifiers

Chemical Name or Material 3-Methylbut-2-enyl formate
Assay Tech.
Molecular Formula C6H10O2
Linear Formula (CH3)2C=CHCH2OCHO
Formula Weight 114.14
MDL Number MFCD00036614
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RDP00092EA

  SDS
fisher scientific company
RDP00092EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

RDP00092DA

  SDS
fisher scientific company
RDP00092DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 114.14
Purity Grade Technical Grade
Quantity 10g
Assay Tech.
Packaging Amber Glass Bottle
Linear Formula (CH3)2C=CHCH2OCHO
Chemical Name or Material 3-Methylbut-2-enyl formate
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
MDL Number MFCD00036614
CAS 68480-28-4
Molecular Formula C6H10O2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.