missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Methylbiphenyl 95.0+%, TCI America™
Supplier: TCI America M02961G
Specifications
3-Methylbiphenyl | |
272°C | |
MFCD00008533 | |
3-methylbiphenyl, 3-phenyltoluene, 3-methyl-1,1'biphenyl, 1,1'-biphenyl, 3-methyl, 3-methyl-1,1'-biphenyl, biphenyl, 3-methyl, unii-kfp6eo4j6p, kfp6eo4j6p, 1-methyl-3-phenyl-benzene, 3-methyl-biphenyl | |
CC1=CC=CC(=C1)C2=CC=CC=C2 | |
168.239 | |
168.24 | |
Liquid |
643-93-6 | |
C13H12 | |
1 g | |
NPDIDUXTRAITDE-UHFFFAOYSA-N | |
1-methyl-3-phenylbenzene | |
12564 | |
≥95.0% (GC) |
Chemical Identifiers
643-93-6 | |
168.239 | |
NPDIDUXTRAITDE-UHFFFAOYSA-N | |
12564 | |
CC1=CC=CC(=C1)C2=CC=CC=C2 |
C13H12 | |
MFCD00008533 | |
3-methylbiphenyl, 3-phenyltoluene, 3-methyl-1,1'biphenyl, 1,1'-biphenyl, 3-methyl, 3-methyl-1,1'-biphenyl, biphenyl, 3-methyl, unii-kfp6eo4j6p, kfp6eo4j6p, 1-methyl-3-phenyl-benzene, 3-methyl-biphenyl | |
1-methyl-3-phenylbenzene |
Safety and Handling
EINECSNumber : (4)-0015
TSCA : Yes