missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3'-Methylbiphenyl-4-sulfonyl chloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC450520010

 View more versions of this product

Catalog No. AC450520010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3'-Methylbiphenyl-4-sulfonylchloride
1g
96% min. (HPLC)
1,1'-biphenyl-4-sulfonylchloride, 3'-methyl, 3'-methyl 1,1'-biphenyl-4-sulfonyl chloride, 3'-methyl-1,1'-biphenyl-4-sulfonyl chloride, 3'-methyl-biphenyl-4-sulfonyl chloride, 3'-methylbiphenyl-4-sulfonyl chloride, 3/'-methylbiphenyl-4-sulfonyl chloride, 4-3-methylphenyl benzenesulfonyl chloride
CC1=CC=CC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl
266.75
266.75
Glass Bottle
186551-47-3
C13H11ClO2S
OHVJVSGIYVAZFD-UHFFFAOYSA-N
4-(3-methylphenyl)benzenesulfonyl chloride
10754271
97%

Chemical Identifiers

186551-47-3
266.75
1,1'-biphenyl-4-sulfonylchloride, 3'-methyl, 3'-methyl 1,1'-biphenyl-4-sulfonyl chloride, 3'-methyl-1,1'-biphenyl-4-sulfonyl chloride, 3'-methyl-biphenyl-4-sulfonyl chloride, 3'-methylbiphenyl-4-sulfonyl chloride, 3/'-methylbiphenyl-4-sulfonyl chloride, 4-3-methylphenyl benzenesulfonyl chloride
4-(3-methylphenyl)benzenesulfonyl chloride
C13H11ClO2S
OHVJVSGIYVAZFD-UHFFFAOYSA-N
10754271
CC1=CC=CC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Danger

SDS Documents