Please login to your online account to display your discounted pricing

3'-Methylbiphenyl-4-sulfonyl chloride, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3'-Methylbiphenyl-4-sulfonyl chloride
Assay Percent Range 97%
Molecular Formula C13H11ClO2S
Formula Weight 266.75
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC450520010

  SDS
thermo fisher scientific
AC450520010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC450520050

  SDS
thermo fisher scientific
AC450520050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 266.75
Quantity 1g
Chemical Name or Material 3'-Methylbiphenyl-4-sulfonyl chloride
Molecular Formula C13H11ClO2S
Assay Percent Range 97%
CAS 186551-47-3