missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methylbenzyl isothiocyanate, 99%

Manufacturer:  Alfa Aesar™ H5489306

 View more versions of this product

Catalog No. AAH5489306



Specifications

Specifications

3-Methylbenzyl isothiocyanate
127°C (2mmHg)
5g
C9H9NS
UN2922
NUXFAFWRXGFTQJ-UHFFFAOYSA-N
1-(isothiocyanatomethyl)-3-methylbenzene
138010
Liquid
1.09 g/mL
Brown
3696-66-0
MFCD00060410
1-isothiocyanatomethyl-3-methyl-benzene
CC1=CC(=CC=C1)CN=C=S
163.238
163.24
99%

Chemical Identifiers

3696-66-0
163.238
NUXFAFWRXGFTQJ-UHFFFAOYSA-N
138010
CC1=CC(=CC=C1)CN=C=S
C9H9NS
MFCD00060410
1-isothiocyanatomethyl-3-methyl-benzene
1-(isothiocyanatomethyl)-3-methylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314-H335

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents