Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methylbenzotrifluoride, 97%

Chemical Identifiers

Chemical Name or Material 3-Methylbenzotrifluoride
CAS 401-79-6
Molecular Formula C8H7F3
Formula Weight 160.14
MDL Number MFCD00190105
UN Number UN1993
Beilstein 1938201
Synonym 3-(Trifluoromethyl)toluene; alpha,alpha,alpha-Trifluoro-m-xylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2001703

  SDS
alfa aesar™
B2001703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2001706

  SDS
alfa aesar™
B2001706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 160.14
Chemical Name or Material 3-Methylbenzotrifluoride
Quantity 1g
Assay 97%
CAS 401-79-6
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
UN Number UN1993
Beilstein 1938201
MDL Number MFCD00190105
Molecular Formula C8H7F3
Synonym 3-(Trifluoromethyl)toluene; alpha,alpha,alpha-Trifluoro-m-xylene
Boiling Point 130°C
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Flash Point 26°C (78°F)
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.