Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methylbenzothiazolium iodide, 98+%

$98.84 - $375.19

Chemical Identifiers

Chemical Name or Material 3-Methylbenzothiazolium iodide
CAS 2786-31-4
Molecular Formula C8H8INS
Formula Weight 277.13
MDL Number MFCD00050297
Beilstein 3917816
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0483718

  SDS
alfa aesar™
L0483718
50g Each for $98.84

AAL0483730

  SDS
alfa aesar™
L0483730
250g Each for $375.19
Description & Specifications

Specifications

Formula Weight 277.13
Chemical Name or Material 3-Methylbenzothiazolium iodide
CAS 2786-31-4
Assay ≥98%
MDL Number MFCD00050297
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Molecular Formula C8H8INS
Beilstein 3917816
Quantity 50g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
TSCA TSCA
Melting Point 208° to 210°C

Applications
3-Methylbenzothiazolium iodide, is used as a pharmaceutical intermediate and also in organic chemical synthesis.

Solubility
Soluble in hot methanol- almost transparency.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.