Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methylbenzophenone, 99%

$181.64 - $800.42

Chemical Identifiers

Chemical Name or Material 3-Methylbenzophenone
CAS 643-65-2
Molecular Formula C14H12O
Formula Weight 196.25
MDL Number MFCD00008535
Beilstein 2045488
Synonym Phenyl m-tolyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1956818

 
alfa aesar™
A1956818
50g Each for $181.64

AAA1956830

 
alfa aesar™
A1956830
250g Each for $800.42
Description & Specifications

Specifications

Formula Weight 196.25
Chemical Name or Material 3-Methylbenzophenone
Assay 99%
Molecular Formula C14H12O
MDL Number MFCD00008535
CAS 643-65-2
Beilstein 2045488
Synonym Phenyl m-tolyl ketone
Flash Point >110°C (230°F)
Boiling Point 316°C
Quantity 50g

Applications
3-Methylbenzophenone is used as a pharmaceutical intermediate or Ketoprofen intermediates.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.