Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methylbenzophenone, 97%

$46.80 - $278.61

Chemical Identifiers

Chemical Name or Material 3-Methylbenzophenone
CAS 643-65-2
Molecular Formula C14H12O
Formula Weight 196.25
MDL Number MFCD00008535
Beilstein 2045488
Synonym Phenyl m-tolyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2566114

 
alfa aesar™
B2566114
25g Each for $46.80

AAB2566122

 
alfa aesar™
B2566122
100g Each for $95.28

AAB2566136

 
alfa aesar™
B2566136
500g Each for $278.61
Description & Specifications

Specifications

Formula Weight 196.25
Chemical Name or Material 3-Methylbenzophenone
Assay 97%
Molecular Formula C14H12O
MDL Number MFCD00008535
CAS 643-65-2
Beilstein 2045488
Synonym Phenyl m-tolyl ketone
Flash Point >110°C (230°F)
Boiling Point 183° to 185°C (16mmHg)
Quantity 25g