missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methylbenzofuran 98.0+%, TCI America™

$153.06 - $495.41

Chemical Identifiers

CAS 21535-97-7
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.162
MDL Number MFCD00205641
InChI Key ZRXHLJNBNWVNIM-UHFFFAOYSA-N
Synonym #, 3-methyl-benzofuran, 3-methylbenzo b furan, 3-methylbenzofuran, acmc-209flj, benzofuran, 3-methyl
PubChem CID 88939
IUPAC Name 3-methyl-1-benzofuran
SMILES CC1=COC2=CC=CC=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M21121G View Documents Tci America
M21121G
1g Each for $153.06
 
M21125G View Documents Tci America
M21125G
5g Each for $495.41
 
Specifications

Chemical Identifiers

21535-97-7
132.162
ZRXHLJNBNWVNIM-UHFFFAOYSA-N
88939
CC1=COC2=CC=CC=C12
C9H8O
MFCD00205641
#, 3-methyl-benzofuran, 3-methylbenzo b furan, 3-methylbenzofuran, acmc-209flj, benzofuran, 3-methyl
3-methyl-1-benzofuran

Specifications

197°C
1g
C9H8O
#, 3-methyl-benzofuran, 3-methylbenzo b furan, 3-methylbenzofuran, acmc-209flj, benzofuran, 3-methyl
CC1=COC2=CC=CC=C12
132.162
132.16
≥98.0% (GC)
Yellow
21535-97-7
MFCD00205641
ZRXHLJNBNWVNIM-UHFFFAOYSA-N
3-methyl-1-benzofuran
88939
Clear Liquid at 20°C
3-Methylbenzofuran
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator