Please login to your online account to display your discounted pricing

3-Methylbenzofuran-2-carbonyl chloride 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Methylbenzofuran-2-carbonyl chloride
Assay Percent Range 97%
Molecular Formula C10H7ClO2
Formula Weight 194.62
MDL Number MFCD01940394
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC372660050

  SDS
thermo fisher scientific
AC372660050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 194.62
Quantity 5g
CAS 2256-86-2
MDL Number MFCD01940394
Assay Percent Range 97%
Chemical Name or Material 3-Methylbenzofuran-2-carbonyl chloride
Molecular Formula C10H7ClO2