Please login to your online account to display your discounted pricing

3-Methylbenzofuran-2-carbonyl chloride 97%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC372660050

 View more versions of this product

Catalog No. AC372660050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 194.62
Quantity 5g
CAS 2256-86-2
MDL Number MFCD01940394
Assay Percent Range 97%
Chemical Name or Material 3-Methylbenzofuran-2-carbonyl chloride
Molecular Formula C10H7ClO2

Specifications

Chemical Name or Material 3-Methylbenzofuran-2-carbonyl chloride
Assay Percent Range 97%
Molecular Formula C10H7ClO2
Formula Weight 194.62
MDL Number MFCD01940394