Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methylbenzo[f]quinoline, 97%

$278.61 - $863.54

Chemical Identifiers

Chemical Name or Material 3-Methylbenzo[f]quinoline
CAS 85-06-3
Molecular Formula C14H11N
Formula Weight 193.25
MDL Number MFCD00046906
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2492014

 
alfa aesar™
B2492014
25g Each for $278.61

AAB2492022

 
alfa aesar™
B2492022
100g Each for $863.54
Description & Specifications

Specifications

Formula Weight 193.25
Chemical Name or Material 3-Methylbenzo[f]quinoline
TSCA TSCA
Assay 97%
Molecular Formula C14H11N
MDL Number MFCD00046906
CAS 85-06-3
Health Hazard 1 Warning
Melting Point 73° to 78°C
Quantity 25g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
3-Methylbenzo[f]quinoline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.