Please login to your online account to display your discounted pricing

3-Methylbenzenethiol, 95%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC421270050

 View more versions of this product

Catalog No. AC421270050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 124.21
Physical Form Liquid
Assay ≥96.0%
Quantity 5g
Color Clear colorless to light yellow
TSCA TSCA
RTECS Number XT8700000
EINECS Number 203-578-4
Density 1.0440g/mL
CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Harmful if inhaled.
Toxic in contact with skin.
Health Hazard 1 GHS Signal Word: Danger
Packaging Glass bottle
Synonym 3-Methylbenzenethiol
Linear Formula CH3C6H4SH
CAS 108-40-7
Molecular Formula C7H8S
Assay Percent Range 95%
Name Note 97%
Chemical Name or Material 3-Methylbenzenethiol

Specifications

Chemical Name or Material 3-Methylbenzenethiol
Name Note 97%
CAS 108-40-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 95%
Molecular Formula C7H8S
Linear Formula CH3C6H4SH
Formula Weight 124.21
Synonym 3-Methylbenzenethiol
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Harmful if inhaled.
Toxic in contact with skin.

SDS & Safety Documents