Please login to your online account to display your discounted pricing

3-Methylbenzene-1,2-diamine, 97%

Chemical Identifiers

Chemical Name or Material 3-Methylbenzene-1, 2-diamine
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H10N2
Linear Formula CH3C6H3(NH2)2
Formula Weight 122.17
Synonym 2, 3-Toluenediamine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BTB07447DA

  SDS
fisher scientific company
BTB07447DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BTB07447EB

  SDS
fisher scientific company
BTB07447EB
Amber Glass Bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BTB07447EA

  SDS
fisher scientific company
BTB07447EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 122.17
Quantity 1g
Assay 97%
Chemical Name or Material 3-Methylbenzene-1, 2-diamine
Packaging Amber Glass Bottle
Assay Percent Range 97%
Linear Formula CH3C6H3(NH2)2
CAS 2687-25-4
Synonym 2, 3-Toluenediamine
Molecular Formula C7H10N2