Please login to your online account to display your discounted pricing

3-Methylbenzene-1,2-diamine, 97%


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB07447DA

 View more versions of this product

Catalog No. BTB07447DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 122.17
Quantity 1g
Assay 97%
CAS 2687-25-4
Chemical Name or Material 3-Methylbenzene-1, 2-diamine
Linear Formula CH3C6H3(NH2)2
Packaging Amber Glass Bottle
Synonym 2, 3-Toluenediamine
Assay Percent Range 97%
Molecular Formula C7H10N2

Specifications

Chemical Name or Material 3-Methylbenzene-1, 2-diamine
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H10N2
Linear Formula CH3C6H3(NH2)2
Formula Weight 122.17
Synonym 2, 3-Toluenediamine