Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methylbenzamidoxime, 96%

$106.08 - $373.68

Chemical Identifiers

Chemical Name or Material 3-Methylbenzamidoxime
CAS 40067-82-1
Molecular Formula C8H10N2O
Formula Weight 150.18
MDL Number MFCD00655170
Synonym N-Hydroxy-3-methylbenzamidine; N-Hydroxy-3-methylbenzenecarboximidamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5068203

  SDS
alfa aesar™
H5068203
1g Each for $106.08

AAH5068206

  SDS
alfa aesar™
H5068206
5g Each for $373.68
Description & Specifications

Specifications

Formula Weight 150.18
Synonym N-Hydroxy-3-methylbenzamidine; N-Hydroxy-3-methylbenzenecarboximidamide
Quantity 1g
Assay 96%
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 87° to 90°C
Molecular Formula C8H10N2O
MDL Number MFCD00655170
CAS 40067-82-1
Chemical Name or Material 3-Methylbenzamidoxime
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.