Learn More
(3-Methylaminopropyl)trimethoxysilane, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1654122
Description
(3-Methylaminopropyl)trimethoxysilane is used as coupling agent, adhesion promoters
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(3-Methylaminopropyl)trimethoxysilane is used as coupling agent, adhesion promoters
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
106°C (30mmHg) | |
3069-25-8 | |
C7H19NO3Si | |
2072720 | |
n-methylaminopropyltrimethoxysilane, n-methyl-3-trimethoxysilyl propan-1-amine, trimethoxy 3-methylamino propyl silane, 1-propanamine, n-methyl-3-trimethoxysilyl, n-methyl-3-trimethoxysilyl propylamine, n-methyl-3-aminopropyltrimethoxysilane, 3-methylaminopropyl-trimethoxysilane, methyl 3-trimethoxysilyl propyl amine, n-methyl-3-aminopropyl-trimethoxysilane, acmc-20ak27 | |
DVYVMJLSUSGYMH-UHFFFAOYSA-N | |
N-methyl-3-trimethoxysilylpropan-1-amine | |
76480 | |
95% | |
Moisture sensitive |
(3-Methylaminopropyl)trimethoxysilane | |
100 g | |
MFCD00048021 | |
82°C (179°F) | |
Hydrolyzes in water. | |
CNCCC[Si](OC)(OC)OC | |
193.32 | |
193.32 | |
1.419 | |
0.978 |
Chemical Identifiers
3069-25-8 | |
193.32 | |
DVYVMJLSUSGYMH-UHFFFAOYSA-N | |
76480 | |
CNCCC[Si](OC)(OC)OC |
C7H19NO3Si | |
MFCD00048021 | |
n-methylaminopropyltrimethoxysilane, n-methyl-3-trimethoxysilyl propan-1-amine, trimethoxy 3-methylamino propyl silane, 1-propanamine, n-methyl-3-trimethoxysilyl, n-methyl-3-trimethoxysilyl propylamine, n-methyl-3-aminopropyltrimethoxysilane, 3-methylaminopropyl-trimethoxysilane, methyl 3-trimethoxysilyl propyl amine, n-methyl-3-aminopropyl-trimethoxysilane, acmc-20ak27 | |
N-methyl-3-trimethoxysilylpropan-1-amine |
Safety and Handling
GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
P210-P280-P305+P351+P338-P310-P370+P378q-P501c
H227-H318
EINECSNumber : 221-334-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only