missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(Methylaminomethyl)pyridine 98.0+%, TCI America™
$74.65 - $242.21
Chemical Identifiers
CAS | 20173-04-0 |
---|---|
Molecular Formula | C7H10N2 |
Molecular Weight (g/mol) | 122.171 |
MDL Number | MFCD00023610 |
InChI Key | MCSAQVGDZLPTBS-UHFFFAOYSA-N |
Synonym | n-methyl-1-pyridin-3-yl methanamine, n-methyl-n-3-pyridylmethyl amine, 3-methylaminomethyl pyridine, n-methyl pyridin-3-yl methanamine, 3-methylamino methyl pyridine, 3-pyridinemethanamine, n-methyl, 3-picolylmethylamine, n-methyl-3-pyridylmethylamine, methyl pyridin-3-ylmethyl amine, n-methyl-3-pyridinemethylamine |
PubChem CID | 88393 |
IUPAC Name | N-methyl-1-pyridin-3-ylmethanamine |
SMILES | CNCC1=CN=CC=C1 |
Chemical Identifiers
20173-04-0 | |
122.171 | |
MCSAQVGDZLPTBS-UHFFFAOYSA-N | |
88393 | |
CNCC1=CN=CC=C1 |
C7H10N2 | |
MFCD00023610 | |
n-methyl-1-pyridin-3-yl methanamine, n-methyl-n-3-pyridylmethyl amine, 3-methylaminomethyl pyridine, n-methyl pyridin-3-yl methanamine, 3-methylamino methyl pyridine, 3-pyridinemethanamine, n-methyl, 3-picolylmethylamine, n-methyl-3-pyridylmethylamine, methyl pyridin-3-ylmethyl amine, n-methyl-3-pyridinemethylamine | |
N-methyl-1-pyridin-3-ylmethanamine |
Specifications
20173-04-0 | |
98°C | |
MFCD00023610 | |
n-methyl-1-pyridin-3-yl methanamine, n-methyl-n-3-pyridylmethyl amine, 3-methylaminomethyl pyridine, n-methyl pyridin-3-yl methanamine, 3-methylamino methyl pyridine, 3-pyridinemethanamine, n-methyl, 3-picolylmethylamine, n-methyl-3-pyridylmethylamine, methyl pyridin-3-ylmethyl amine, n-methyl-3-pyridinemethylamine | |
CNCC1=CN=CC=C1 | |
122.171 | |
122.17 | |
Liquid |
Yellow | |
C7H10N2 | |
1 g | |
MCSAQVGDZLPTBS-UHFFFAOYSA-N | |
N-methyl-1-pyridin-3-ylmethanamine | |
88393 | |
≥98.0% (GC,T) | |
3-(Methylaminomethyl)pyridine |
Safety and Handling
TSCA : No