missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(Methylaminomethyl)pyridine 98.0+%, TCI America™
Supplier: TCI America M22575G
Specifications
3-(Methylaminomethyl)pyridine | |
Yellow | |
C7H10N2 | |
5 g | |
MCSAQVGDZLPTBS-UHFFFAOYSA-N | |
N-methyl-1-pyridin-3-ylmethanamine | |
88393 | |
≥98.0% (GC,T) |
20173-04-0 | |
98°C | |
MFCD00023610 | |
n-methyl-1-pyridin-3-yl methanamine, n-methyl-n-3-pyridylmethyl amine, 3-methylaminomethyl pyridine, n-methyl pyridin-3-yl methanamine, 3-methylamino methyl pyridine, 3-pyridinemethanamine, n-methyl, 3-picolylmethylamine, n-methyl-3-pyridylmethylamine, methyl pyridin-3-ylmethyl amine, n-methyl-3-pyridinemethylamine | |
CNCC1=CN=CC=C1 | |
122.171 | |
122.17 | |
Liquid |
Chemical Identifiers
20173-04-0 | |
122.171 | |
MCSAQVGDZLPTBS-UHFFFAOYSA-N | |
88393 | |
CNCC1=CN=CC=C1 |
C7H10N2 | |
MFCD00023610 | |
n-methyl-1-pyridin-3-yl methanamine, n-methyl-n-3-pyridylmethyl amine, 3-methylaminomethyl pyridine, n-methyl pyridin-3-yl methanamine, 3-methylamino methyl pyridine, 3-pyridinemethanamine, n-methyl, 3-picolylmethylamine, n-methyl-3-pyridylmethylamine, methyl pyridin-3-ylmethyl amine, n-methyl-3-pyridinemethylamine | |
N-methyl-1-pyridin-3-ylmethanamine |
Safety and Handling
TSCA : No