missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Methylaminomethyl)benzonitrile, TCI America™

$201.67 - $605.02

Chemical Identifiers

CAS 90389-96-1
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.193
MDL Number MFCD09731650
InChI Key KBRCDVSFCMVQSN-UHFFFAOYSA-N
Synonym 3-methylamino methyl benzonitrile, 3-methylamino methyl benzonitrile, 3-methylaminomethyl benzonitrile, 3-methylaminomethyl-benzonitrile, benzonitrile, 3-methylamino methyl, benzonitrile, 3-methylamino methyl, 3-methylamino methyl-benzonitrile, 3-cyano-n-methylbenzylamine, n-methyl-n-3-cyanobenzyl amine
PubChem CID 457595
IUPAC Name 3-(methylaminomethyl)benzonitrile
SMILES CNCC1=CC=CC(=C1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M28691G View Documents TCI America
M28691G
1g Each for $201.67
 
M28695G View Documents TCI America
M28695G
5g Each for $605.02
 
Specifications

Chemical Identifiers

90389-96-1
146.193
KBRCDVSFCMVQSN-UHFFFAOYSA-N
457595
CNCC1=CC=CC(=C1)C#N
C9H10N2
MFCD09731650
3-methylamino methyl benzonitrile, 3-methylamino methyl benzonitrile, 3-methylaminomethyl benzonitrile, 3-methylaminomethyl-benzonitrile, benzonitrile, 3-methylamino methyl, benzonitrile, 3-methylamino methyl, 3-methylamino methyl-benzonitrile, 3-cyano-n-methylbenzylamine, n-methyl-n-3-cyanobenzyl amine
3-(methylaminomethyl)benzonitrile

Specifications

145°C
1g
C9H10N2
2922
KBRCDVSFCMVQSN-UHFFFAOYSA-N
3-(methylaminomethyl)benzonitrile
457595
Clear Liquid at 20°C
3-(Methylaminomethyl)benzonitrile
Yellow
90389-96-1
MFCD09731650
3-methylamino methyl benzonitrile, 3-methylamino methyl benzonitrile, 3-methylaminomethyl benzonitrile, 3-methylaminomethyl-benzonitrile, benzonitrile, 3-methylamino methyl, benzonitrile, 3-methylamino methyl, 3-methylamino methyl-benzonitrile, 3-cyano-n-methylbenzylamine, n-methyl-n-3-cyanobenzyl amine
CNCC1=CC=CC(=C1)C#N
146.193
146.19
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No