missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Methylaminomethyl)benzonitrile, 95%

$134.31 - $400.95

Chemical Identifiers

CAS 90389-96-1
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.193
MDL Number MFCD09731650
InChI Key KBRCDVSFCMVQSN-UHFFFAOYSA-N
Synonym 3-methylamino methyl benzonitrile, 3-methylamino methyl benzonitrile, 3-methylaminomethyl benzonitrile, 3-methylaminomethyl-benzonitrile, benzonitrile, 3-methylamino methyl, benzonitrile, 3-methylamino methyl, 3-methylamino methyl-benzonitrile, 3-cyano-n-methylbenzylamine, n-methyl-n-3-cyanobenzyl amine
PubChem CID 457595
IUPAC Name 3-(methylaminomethyl)benzonitrile
SMILES CNCC1=CC=CC(=C1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63054MD View Documents Alfa Aesar™
H63054MD
250mg Each for $134.31
 
AAH6305403 View Documents Alfa Aesar™
H6305403
1g Each for $400.95
 
Description

Description

Applications
3-(Methylaminomethyl)benzonitrile is used as pharmaceutical intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
Specifications

Chemical Identifiers

90389-96-1
146.193
KBRCDVSFCMVQSN-UHFFFAOYSA-N
457595
CNCC1=CC=CC(=C1)C#N
C9H10N2
MFCD09731650
3-methylamino methyl benzonitrile, 3-methylamino methyl benzonitrile, 3-methylaminomethyl benzonitrile, 3-methylaminomethyl-benzonitrile, benzonitrile, 3-methylamino methyl, benzonitrile, 3-methylamino methyl, 3-methylamino methyl-benzonitrile, 3-cyano-n-methylbenzylamine, n-methyl-n-3-cyanobenzyl amine
3-(methylaminomethyl)benzonitrile

Specifications

250mg
C9H10N2
UN3276
KBRCDVSFCMVQSN-UHFFFAOYSA-N
3-(methylaminomethyl)benzonitrile
457595
95%
90389-96-1
MFCD09731650
3-methylamino methyl benzonitrile, 3-methylamino methyl benzonitrile, 3-methylaminomethyl benzonitrile, 3-methylaminomethyl-benzonitrile, benzonitrile, 3-methylamino methyl, benzonitrile, 3-methylamino methyl, 3-methylamino methyl-benzonitrile, 3-cyano-n-methylbenzylamine, n-methyl-n-3-cyanobenzyl amine
CNCC1=CC=CC(=C1)C#N
146.193
146.19
3-(Methylaminomethyl)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

Recommended Storage : −30° to −10°C