missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Methylamino)pyrrolidine, TCI America™

$394.17 - $1,268.08

Chemical Identifiers

CAS 83030-08-4
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.165
MDL Number MFCD00191460
InChI Key NGZYRKGJWYJGRS-UHFFFAOYSA-N
PubChem CID 11788268
IUPAC Name N-methylpyrrolidin-3-amine
SMILES CNC1CCNC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M11065G View Documents TCI America
M11065G
5g Each for $394.17
 
M110625G View Documents TCI America
M110625G
25g Each for $1,268.08
 
Specifications

Chemical Identifiers

83030-08-4
100.165
NGZYRKGJWYJGRS-UHFFFAOYSA-N
N-methylpyrrolidin-3-amine
C5H12N2
MFCD00191460
11788268
CNC1CCNC1

Specifications

165°C
5g
C5H12N2
2734
CNC1CCNC1
100.165
100.17
≥98.0% (GC,T)
Colorless
83030-08-4
MFCD00191460
NGZYRKGJWYJGRS-UHFFFAOYSA-N
N-methylpyrrolidin-3-amine
11788268
Clear Liquid at 20°C
3-(Methylamino)pyrrolidine
Safety and Handling

Safety and Handling

TSCA : Yes