missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Methylamino)pyridine, TCI America™

$68.75 - $232.23

Chemical Identifiers

CAS 18364-47-1
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD01418130
InChI Key DBGFNLVRAFYZBI-UHFFFAOYSA-N
Synonym n-methyl-3-pyridinamine, 3-methylamino pyridine, methyl-pyridin-3-yl-amine, 3-pyridinamine, n-methyl, 3-methylaminopyridine, unii-33vnv59xc6, methylpyridin-3-ylamine, pubchem14983, acmc-1bqbo, pyridin-3-yl-methylamine
PubChem CID 140376
IUPAC Name N-methylpyridin-3-amine
SMILES CNC1=CN=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M26845G View Documents TCI America
M26845G
5g Each for $232.23
 
M26841G View Documents TCI America
M26841G
1g Each for $68.75
 
Specifications

Chemical Identifiers

18364-47-1
108.144
DBGFNLVRAFYZBI-UHFFFAOYSA-N
140376
CNC1=CN=CC=C1
C6H8N2
MFCD01418130
n-methyl-3-pyridinamine, 3-methylamino pyridine, methyl-pyridin-3-yl-amine, 3-pyridinamine, n-methyl, 3-methylaminopyridine, unii-33vnv59xc6, methylpyridin-3-ylamine, pubchem14983, acmc-1bqbo, pyridin-3-yl-methylamine
N-methylpyridin-3-amine

Specifications

110°C
5g
C6H8N2
n-methyl-3-pyridinamine, 3-methylamino pyridine, methyl-pyridin-3-yl-amine, 3-pyridinamine, n-methyl, 3-methylaminopyridine, unii-33vnv59xc6, methylpyridin-3-ylamine, pubchem14983, acmc-1bqbo, pyridin-3-yl-methylamine
CNC1=CN=CC=C1
108.144
108.14
≥98.0% (GC,T)
Undesignated
18364-47-1
MFCD01418130
DBGFNLVRAFYZBI-UHFFFAOYSA-N
N-methylpyridin-3-amine
140376
Clear Liquid at 20°C
3-(Methylamino)pyridine
Safety and Handling

Safety and Handling

TSCA : No