missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Methylamino)pyridine, 95%

Manufacturer:  Alfa Aesar™ H6360203

 View more versions of this product

Catalog No. AAH6360203



Specifications

Specifications

3-(Methylamino)pyridine
1g
C6H8N2
UN2810
DBGFNLVRAFYZBI-UHFFFAOYSA-N
N-methylpyridin-3-amine
140376
Liquid
Yellow
18364-47-1
MFCD01418130
n-methyl-3-pyridinamine, 3-methylamino pyridine, methyl-pyridin-3-yl-amine, 3-pyridinamine, n-methyl, 3-methylaminopyridine, unii-33vnv59xc6, methylpyridin-3-ylamine, pubchem14983, acmc-1bqbo, pyridin-3-yl-methylamine
CNC1=CN=CC=C1
108.144
108.14
95%

Chemical Identifiers

18364-47-1
108.144
DBGFNLVRAFYZBI-UHFFFAOYSA-N
140376
CNC1=CN=CC=C1
C6H8N2
MFCD01418130
n-methyl-3-pyridinamine, 3-methylamino pyridine, methyl-pyridin-3-yl-amine, 3-pyridinamine, n-methyl, 3-methylaminopyridine, unii-33vnv59xc6, methylpyridin-3-ylamine, pubchem14983, acmc-1bqbo, pyridin-3-yl-methylamine
N-methylpyridin-3-amine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

SDS Documents