missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Methylamino)pyridine, 95%

$47.49 - $562.88

Chemical Identifiers

CAS 18364-47-1
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD01418130
InChI Key DBGFNLVRAFYZBI-UHFFFAOYSA-N
Synonym n-methyl-3-pyridinamine, 3-methylamino pyridine, methyl-pyridin-3-yl-amine, 3-pyridinamine, n-methyl, 3-methylaminopyridine, unii-33vnv59xc6, methylpyridin-3-ylamine, pubchem14983, acmc-1bqbo, pyridin-3-yl-methylamine
PubChem CID 140376
IUPAC Name N-methylpyridin-3-amine
SMILES CNC1=CN=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63602MD View Documents Alfa Aesar™
H63602MD
250mg Each for $47.49
 
AAH6360203 View Documents Alfa Aesar™
H6360203
1g Each for $140.80
 
AAH6360206 View Documents Alfa Aesar™
H6360206
5g Each for $562.88
 
Specifications

Chemical Identifiers

18364-47-1
108.144
DBGFNLVRAFYZBI-UHFFFAOYSA-N
140376
CNC1=CN=CC=C1
C6H8N2
MFCD01418130
n-methyl-3-pyridinamine, 3-methylamino pyridine, methyl-pyridin-3-yl-amine, 3-pyridinamine, n-methyl, 3-methylaminopyridine, unii-33vnv59xc6, methylpyridin-3-ylamine, pubchem14983, acmc-1bqbo, pyridin-3-yl-methylamine
N-methylpyridin-3-amine

Specifications

Yellow
18364-47-1
MFCD01418130
n-methyl-3-pyridinamine, 3-methylamino pyridine, methyl-pyridin-3-yl-amine, 3-pyridinamine, n-methyl, 3-methylaminopyridine, unii-33vnv59xc6, methylpyridin-3-ylamine, pubchem14983, acmc-1bqbo, pyridin-3-yl-methylamine
CNC1=CN=CC=C1
108.144
108.14
95%
250mg
C6H8N2
UN2810
DBGFNLVRAFYZBI-UHFFFAOYSA-N
N-methylpyridin-3-amine
140376
Liquid
3-(Methylamino)pyridine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.