Please login to your online account to display your discounted pricing

3-Methylamino-1,2-propanediol, 98%, ACROS Organics™


Quantity 100mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  35253-1000

 View more versions of this product

Catalog No. AC352531000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Viscous Liquid
Formula Weight 105.14
Quantity 100mL
Color Clear colorless to pale yellow
EINECS Number 254-809-0
TSCA TSCA
Density 1.09g/mL
CAS Max % 100.0
Chemical Name or Material 3-Methylamino-1, 2-propanediol
Refractive Index 1.4740 to 1.4780 (20°C, 589nm)
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C4H11NO2
Linear Formula H3CNHCHCH(OH)CHOH
MDL Number MFCD00082732
CAS 40137-22-2
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 3-Methylamino-1, 2-propanediol
Name Note 98%
CAS 40137-22-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H11NO2
Linear Formula H3CNHCHCH(OH)CHOH
Formula Weight 105.14
MDL Number MFCD00082732
Accessories

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.

SDS & Safety Documents