Please login to your online account to display your discounted pricing

3-Methylamino-1,2-propanediol, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Methylamino-1, 2-propanediol
Name Note 98%
CAS 40137-22-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H11NO2
Linear Formula H3CNHCHCH(OH)CHOH
Formula Weight 105.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC352531000

  SDS
thermo fisher scientific
35253-1000
Glass bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 105.14
Physical Form Viscous Liquid
Quantity 100mL
Color Clear colorless to pale yellow
TSCA TSCA
EINECS Number 254-809-0
Density 1.09g/mL
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Refractive Index 1.4740 to 1.4780 (20°C, 589nm)
Infrared Spectrum Authentic
Name Note 98%
Molecular Formula C4H11NO2
Chemical Name or Material 3-Methylamino-1, 2-propanediol
Assay Percent Range 98%
Linear Formula H3CNHCHCH(OH)CHOH
MDL Number MFCD00082732
CAS 40137-22-2