Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyladipic acid, 99%

$106.35 - $344.80

Chemical Identifiers

Chemical Name or Material 3-Methyladipic acid
CAS 3058-01-3
Molecular Formula HO2CCH2CH(CH3)CH2CH2CO2H
Formula Weight 160.17
MDL Number MFCD00002804
Synonym 3-Methylhexanedioic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2138714

 
alfa aesar™
B2138714
25g Each for $106.35

AAB2138722

 
alfa aesar™
B2138722
100g Each for $344.80
Description & Specifications

Specifications

Formula Weight 160.17
Chemical Name or Material 3-Methyladipic acid
Assay 99%
Molecular Formula HO2CCH2CH(CH3)CH2CH2CO2H
MDL Number MFCD00002804
Boiling Point 229° to 231°C (30mmHg)
CAS 3058-01-3
Melting Point 97° to 102°C
Synonym 3-Methylhexanedioic acid
Quantity 25g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.