missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3′-Methylacetophenone 97.0+%, TCI America™

Manufacturer:  Tci America M0191100ML

 View more versions of this product

Catalog No. M0191100ML



Specifications

Specifications

3′-Methylacetophenone
Yellow
585-74-0
MFCD00008742
FSPSELPMWGWDRY-UHFFFAOYSA-N
1-(3-methylphenyl)ethanone
11455
Clear Liquid at 20°C
219°C
100mL
C9H10O
1-3-methylphenyl ethan-1-one, 1-m-tolyl ethanone, 3'-methylacetophenone, 3-acetyltoluene, 3-methylacetophenone, acetophenone, 3'-methyl, acetophenone, m-methyl, ethanone, 1-3-methylphenyl, m-methylacetophenone, methyl m-tolyl ketone
CC1=CC=CC(=C1)C(=O)C
134.178
134.18
≥97.0% (GC)

Chemical Identifiers

585-74-0
134.178
FSPSELPMWGWDRY-UHFFFAOYSA-N
11455
CC1=CC=CC(=C1)C(=O)C
C9H10O
MFCD00008742
1-3-methylphenyl ethan-1-one, 1-m-tolyl ethanone, 3'-methylacetophenone, 3-acetyltoluene, 3-methylacetophenone, acetophenone, 3'-methyl, acetophenone, m-methyl, ethanone, 1-3-methylphenyl, m-methylacetophenone, methyl m-tolyl ketone
1-(3-methylphenyl)ethanone
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1239

TSCA : No

Documents