Please login to your online account to display your discounted pricing

3'-Methylacetophenone, 97%, ACROS Organics™

$33.78 - $33.78

Chemical Identifiers

Chemical Name or Material 3';-Methylacetophenone
CAS 585-74-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H10O
Linear Formula CH3C6H4COCH3
Formula Weight 134.18
Synonym 3-Acetyltoluene
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC126170250

  SDS
thermo fisher scientific
AC126170250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC126170050

  SDS
thermo fisher scientific
AC126170050
Glass bottle 5g Each for $33.78

AC126171000

  SDS
thermo fisher scientific
AC126171000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 134.18
Quantity 25g
Color Clear colorless to slightly yellow
Density 1.0100g/mL
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Molecular Formula C9H10O
Linear Formula CH3C6H4COCH3
Synonym 3-Acetyltoluene
Health Hazard 1 Warning
Packaging Glass bottle
Chemical Name or Material 3';-Methylacetophenone
Assay Percent Range 97%
CAS 585-74-0
Safety and Handling

Warning