missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3'-Methylacetophenone, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC126171000

 View more versions of this product

Catalog No. AC126171000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

3';-Methylacetophenone
96.0
Glass bottle
Liquid
97%
C9H10O
134.18
3-Acetyltoluene
585-74-0
100.0
Authentic
≥96.0%
1.0100g/mL
CH3C6H4COCH3
Clear colorless to slightly yellow
100g

Chemical Identifiers

585-74-0
100.0
C9H10O
134.18
3';-Methylacetophenone
96.0
97%
CH3C6H4COCH3
3-Acetyltoluene
≥96.0%

Safety and Handling

SDS & Safety Documents

Warning