Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl(thiobenzamide), 97%

$85.43 - $260.68

Chemical Identifiers

Chemical Name or Material 3-Methyl(thiobenzamide)
CAS 2362-63-2
Molecular Formula C8H9NS
Formula Weight 151.23
MDL Number MFCD01314035
Synonym 3-Methylbenzene-1-carbothioamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2646803

  SDS
alfa aesar™
H2646803
1g Each for $85.43

AAH2646806

  SDS
alfa aesar™
H2646806
5g Each for $260.68
Description & Specifications

Specifications

Formula Weight 151.23
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Quantity 1g
Health Hazard 1 Warning
Molecular Formula C8H9NS
MDL Number MFCD01314035
Assay 97%
CAS 2362-63-2
Synonym 3-Methylbenzene-1-carbothioamide
Health Hazard 3 GHS P Statement
P280f
Wear protective clothing.
Melting Point 87° to 89°C
Chemical Name or Material 3-Methyl(thiobenzamide)
Safety and Handling

Warning

GHS P Statement
P280f
Wear protective clothing.

GHS H Statement
H302
Harmful if swallowed.