Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-N-methylbenzylamine, 97%

Chemical Identifiers

Chemical Name or Material 3-Methyl-N-methylbenzylamine
CAS 39180-84-2
Molecular Formula C9H13N
Formula Weight 135.21
MDL Number MFCD05865181
UN Number UN3259
Synonym 3-Methylbenzylmethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6323103

  SDS
alfa aesar™
H6323103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6323106

  SDS
alfa aesar™
H6323106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 135.21
Boiling Point 90° to 92°C (15mmHg)
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
CAS 39180-84-2
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Quantity 1g
Assay 97%
Flash Point 78°C (172°F)
Synonym 3-Methylbenzylmethylamine
Health Hazard 1 Danger
UN Number UN3259
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C9H13N
MDL Number MFCD05865181
Chemical Name or Material 3-Methyl-N-methylbenzylamine

Applications
3-Methyl-N-methylbenzylamine, is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.