Please login to your online account to display your discounted pricing

3-Methyl-GABA, Tocris


Activator of GABA amino-transferase

Quantity 10mg

Manufacturer: r & d systems  RDS038610

 View more versions of this product

Catalog No. 038610

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 522.58
Chemical Name or Material 3-Methyl-GABA
Molecular Formula C5H11NO2
Assay Percent Range >98%
Synonym (±)-4-Amino-3-methylbutanoic acid hemi naphthalene-1, 5-disulfonic acid
CAS 71424-95-8
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material 3-Methyl-GABA
Assay Percent Range >98%
Molecular Formula C5H11NO2
Formula Weight 522.58
Synonym (±)-4-Amino-3-methylbutanoic acid hemi naphthalene-1, 5-disulfonic acid
Accessories

Store at Room Temperature