Please login to your online account to display your discounted pricing

3-Methyl-GABA, Tocris

Activator of GABA amino-transferase

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material 3-Methyl-GABA
Assay Percent Range >98%
Molecular Formula C5H11NO2
Formula Weight 522.58
Synonym (±)-4-Amino-3-methylbutanoic acid hemi naphthalene-1, 5-disulfonic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

038610

 
r & d systems
RDS038610
10mg Each for $164.61

038650

 
r & d systems
RDS038650
50mg Each for $682.67
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 522.58
Chemical Name or Material 3-Methyl-GABA
Molecular Formula C5H11NO2
Assay Percent Range >98%
Synonym (±)-4-Amino-3-methylbutanoic acid hemi naphthalene-1, 5-disulfonic acid
CAS 71424-95-8
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature