Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-D-phenylalanine, 98%

$156.70 - $156.70

Chemical Identifiers

Chemical Name or Material 3-Methyl-D-phenylalanine
CAS 114926-39-5
Molecular Formula C10H13NO2
Formula Weight 179.22
MDL Number MFCD01860871
Synonym H-D-Phe(3-Me)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27108MD

  SDS
alfa aesar™
H27108MD
250mg Each for $156.70
Description & Specifications

Specifications

Formula Weight 179.22
Chemical Name or Material 3-Methyl-D-phenylalanine
Assay 98%
CAS 114926-39-5
Synonym H-D-Phe(3-Me)-OH
Quantity 250mg
Molecular Formula C10H13NO2
MDL Number MFCD01860871