Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-beta-styrylboronic acid pinacol ester, 97%

Chemical Identifiers

Chemical Name or Material 3-Methyl-beta-styrylboronic acid pinacol ester
Molecular Formula C15H21BO2
Formula Weight 244.14
MDL Number MFCD12546181
Synonym 4,4,5,5-Tetramethyl-2-(trans-2-m-tolylvinyl)[1,3,2]dioxaborolane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3007303

  SDS
alfa aesar™
H3007303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3007306

  SDS
alfa aesar™
H3007306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3007314

  SDS
alfa aesar™
H3007314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 244.14
Chemical Name or Material 3-Methyl-beta-styrylboronic acid pinacol ester
Assay 97%
Molecular Formula C15H21BO2
MDL Number MFCD12546181
Synonym 4,4,5,5-Tetramethyl-2-(trans-2-m-tolylvinyl)[1,3,2]dioxaborolane
Boiling Point 114°C (0.3mmHg)
Quantity 1g