Please login to your online account to display your discounted pricing

3-Methyl-5-(trifluoromethyl)pyrazole, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC398600010

 View more versions of this product

Catalog No. AC398600010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥98.5%
Formula Weight 150.1
Quantity 1g
Color Off-white
Melting Point 88° to 92°C
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3-Methyl-5-(trifluoromethyl)pyrazole
Name Note 99%
Assay Percent Range 99%
Molecular Formula C5H5F3N2
MDL Number MFCD00829309
CAS 10010-93-2
CAS Min % 98.5
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

Specifications

Chemical Name or Material 3-Methyl-5-(trifluoromethyl)pyrazole
Name Note 99%
CAS 10010-93-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C5H5F3N2
Formula Weight 150.1
MDL Number MFCD00829309
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents