missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Methyl-5-(trifluoromethyl)pyrazole, 99%, ACROS Organics™
Chemical Identifiers
CAS | 10010-93-2 |
---|---|
Molecular Formula | C5H5F3N2 |
Molecular Weight (g/mol) | 150.1 |
MDL Number | MFCD00829309 |
InChI Key | DLCHCAYDSKIFIN-UHFFFAOYSA-N |
Synonym | 1h-pyrazole, 3-methyl-5-trifluoromethyl, 3-methyl-5-trifluoromethyl pyrazole, 3-methyl-5-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl-2h-pyrazole, 3-methyl-5-trifluoromethylpyrazole, 3-trifluoromethyl-5-methyl pyrazole, 3-trifluoromethyl-5-methylpyrazole, 5-methyl-3-trifluoromethyl pyrazole, 5-methyl-3-trifluoromethyl-1h-pyrazole, pyrazole, 3-methyl-5-trifluoromethyl |
PubChem CID | 139077 |
IUPAC Name | 5-methyl-3-(trifluoromethyl)-1H-pyrazole |
SMILES | CC1=CC(=NN1)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC398600010
|
Thermo Fisher Scientific
AC398600010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC398600050
|
Thermo Fisher Scientific
AC398600050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
10010-93-2 | |
150.1 | |
DLCHCAYDSKIFIN-UHFFFAOYSA-N | |
139077 | |
CC1=CC(=NN1)C(F)(F)F |
C5H5F3N2 | |
MFCD00829309 | |
1h-pyrazole, 3-methyl-5-trifluoromethyl, 3-methyl-5-trifluoromethyl pyrazole, 3-methyl-5-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl-2h-pyrazole, 3-methyl-5-trifluoromethylpyrazole, 3-trifluoromethyl-5-methyl pyrazole, 3-trifluoromethyl-5-methylpyrazole, 5-methyl-3-trifluoromethyl pyrazole, 5-methyl-3-trifluoromethyl-1h-pyrazole, pyrazole, 3-methyl-5-trifluoromethyl | |
5-methyl-3-(trifluoromethyl)-1H-pyrazole |
Specifications
Authentic | |
White | |
1g | |
98.5 | |
98.5% min. (GC) | |
MFCD00829309 | |
DLCHCAYDSKIFIN-UHFFFAOYSA-N | |
5-methyl-3-(trifluoromethyl)-1H-pyrazole | |
139077 | |
Powder | |
3-Methyl-5-(trifluoromethyl)pyrazole, 99% |
Glass bottle | |
88.0°C to 90.0°C | |
10010-93-2 | |
100.0 | |
C5H5F3N2 | |
1h-pyrazole, 3-methyl-5-trifluoromethyl, 3-methyl-5-trifluoromethyl pyrazole, 3-methyl-5-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl-2h-pyrazole, 3-methyl-5-trifluoromethylpyrazole, 3-trifluoromethyl-5-methyl pyrazole, 3-trifluoromethyl-5-methylpyrazole, 5-methyl-3-trifluoromethyl pyrazole, 5-methyl-3-trifluoromethyl-1h-pyrazole, pyrazole, 3-methyl-5-trifluoromethyl | |
CC1=CC(=NN1)C(F)(F)F | |
150.1 | |
150.1 | |
99% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning