missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-5-(trifluoromethyl)benzoyl chloride, 97%

Manufacturer:  Alfa Aesar™ H3318203

 View more versions of this product

Catalog No. AAH3318203



Specifications

Specifications

3-Methyl-5-(trifluoromethyl)benzoyl chloride
1g
C9H6ClF3O
UN3265
STYLTOJEHPKJDF-UHFFFAOYSA-N
3-methyl-5-(trifluoromethyl)benzoyl chloride
2775605
Liquid
Moisture Sensitive
261952-09-4
MFCD1631600
3-methyl-5-trifluoromethyl benzoyl chloride, 5-methyl-3-trifluoromethyl benzoyl chloride
CC1=CC(=CC(=C1)C(=O)Cl)C(F)(F)F
222.591
222.59
97%

Chemical Identifiers

261952-09-4
222.591
STYLTOJEHPKJDF-UHFFFAOYSA-N
2775605
CC1=CC(=CC(=C1)C(=O)Cl)C(F)(F)F
C9H6ClF3O
MFCD1631600
3-methyl-5-trifluoromethyl benzoyl chloride, 5-methyl-3-trifluoromethyl benzoyl chloride
3-methyl-5-(trifluoromethyl)benzoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

SDS Documents