missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Methyl-5-phenyl-1H-pyrazole, Thermo Scientific Chemicals
$233.55 - $233.55
Chemical Identifiers
CAS | 3347-62-4 |
---|---|
Molecular Formula | C10H10N2 |
Molecular Weight (g/mol) | 158.20 |
MDL Number | MFCD00022385,MFCD00524752 |
InChI Key | QHRSESMSOJZMCO-UHFFFAOYSA-N |
PubChem CID | 18774 |
IUPAC Name | 5-methyl-3-phenyl-1H-pyrazole |
SMILES | CC1=CC(=NN1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5065003
|
Thermo Scientific Chemicals
H5065003 |
1 g |
Each for $233.55
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3347-62-4 | |
158.20 | |
QHRSESMSOJZMCO-UHFFFAOYSA-N | |
5-methyl-3-phenyl-1H-pyrazole |
C10H10N2 | |
MFCD00022385,MFCD00524752 | |
18774 | |
CC1=CC(=NN1)C1=CC=CC=C1 |
Specifications
3347-62-4 | |
C10H10N2 | |
1 g | |
QHRSESMSOJZMCO-UHFFFAOYSA-N | |
5-methyl-3-phenyl-1H-pyrazole | |
18774 | |
3-Methyl-5-phenyl-1H-pyrazole |
124°C to 127°C | |
MFCD00022385,MFCD00524752 | |
3-methyl-5-phenyl-1h-pyrazole, femerazol, 3-methyl-5-phenylpyrazole, phemerazole, 1h-pyrazole, 3-methyl-5-phenyl, 5-methyl-3-phenylpyrazole, 1h-pyrazole,3-methyl-5-phenyl, pyrazole, 3-methyl-5-phenyl, unii-0xfx19d77a, 1h-pyrazole, 5-methyl-3-phenyl | |
CC1=CC(=NN1)C1=CC=CC=C1 | |
158.20 | |
158.2 |
Safety and Handling
EINECSNumber : 222-099-1
RTECSNumber : UQ7525000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only