Learn More
3-Methyl-5-nitropyridine, 97%, Thermo Scientific Chemicals
$519.85 - $519.85
Chemical Identifiers
CAS | 6960-20-9 |
---|---|
Molecular Formula | C6H6N2O2 |
Molecular Weight (g/mol) | 138.126 |
MDL Number | MFCD04114214 |
InChI Key | OQWXZZCTJZOSGH-UHFFFAOYSA-N |
Synonym | 3-nitro-5-picoline, 5-nitro-3-picoline, pyridine,3-methyl-5-nitro, pubchem6645, 5-methyl-3-nitropyridine, 3-nitro-5-methylpyridine, acmc-209o9m, 3-methyl-5-nitro-pyridine, pyridine, 3-methyl-5-nitro, pyridine, 3-methyl-5-nitro-9ci |
PubChem CID | 250373 |
IUPAC Name | 3-methyl-5-nitropyridine |
SMILES | CC1=CC(=CN=C1)[N+](=O)[O-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH3168306
|
Thermo Scientific Chemicals
H3168306 |
5 g |
Each for $519.85
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6960-20-9 | |
138.126 | |
OQWXZZCTJZOSGH-UHFFFAOYSA-N | |
250373 | |
CC1=CC(=CN=C1)[N+](=O)[O-] |
C6H6N2O2 | |
MFCD04114214 | |
3-nitro-5-picoline, 5-nitro-3-picoline, pyridine,3-methyl-5-nitro, pubchem6645, 5-methyl-3-nitropyridine, 3-nitro-5-methylpyridine, acmc-209o9m, 3-methyl-5-nitro-pyridine, pyridine, 3-methyl-5-nitro, pyridine, 3-methyl-5-nitro-9ci | |
3-methyl-5-nitropyridine |
Specifications
6960-20-9 | |
C6H6N2O2 | |
5 g | |
3-nitro-5-picoline, 5-nitro-3-picoline, pyridine,3-methyl-5-nitro, pubchem6645, 5-methyl-3-nitropyridine, 3-nitro-5-methylpyridine, acmc-209o9m, 3-methyl-5-nitro-pyridine, pyridine, 3-methyl-5-nitro, pyridine, 3-methyl-5-nitro-9ci | |
CC1=CC(=CN=C1)[N+](=O)[O-] | |
138.126 | |
138.13 | |
3-Methyl-5-nitropyridine |
94°C to 98°C | |
MFCD04114214 | |
UN2811 | |
OQWXZZCTJZOSGH-UHFFFAOYSA-N | |
3-methyl-5-nitropyridine | |
250373 | |
97% |
Safety and Handling
GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
H302+H332-H311-H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only