missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-4-nitrothiobenzamide, 97%

$89.35 - $1,179.59

Chemical Identifiers

CAS 886364-22-3
Molecular Formula C8H8N2O2S
Molecular Weight (g/mol) 196.224
MDL Number MFCD06738330
InChI Key QNPFIXDGSZDAGC-UHFFFAOYSA-N
Synonym 3-methyl-4-nitrobenzene-1-carbothioamide
PubChem CID 55255791
IUPAC Name 3-methyl-4-nitrobenzenecarbothioamide
SMILES CC1=C(C=CC(=C1)C(=S)N)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54550MD View Documents Alfa Aesar™
H54550MD
250mg Each for $89.35
 
AAH5455003 View Documents Alfa Aesar™
H5455003
1g Each for $282.69
 
AAH5455006 View Documents Alfa Aesar™
H5455006
5g Each for $1,179.59
 
Specifications

Chemical Identifiers

886364-22-3
196.224
QNPFIXDGSZDAGC-UHFFFAOYSA-N
55255791
CC1=C(C=CC(=C1)C(=S)N)[N+](=O)[O-]
C8H8N2O2S
MFCD06738330
3-methyl-4-nitrobenzene-1-carbothioamide
3-methyl-4-nitrobenzenecarbothioamide

Specifications

250mg
C8H8N2O2S
19566115
QNPFIXDGSZDAGC-UHFFFAOYSA-N
3-methyl-4-nitrobenzenecarbothioamide
55255791
97%
886364-22-3
MFCD06738330
3-methyl-4-nitrobenzene-1-carbothioamide
CC1=C(C=CC(=C1)C(=S)N)[N+](=O)[O-]
196.224
196.22
3-Methyl-4-nitrothiobenzamide
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures