missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-4-nitrothiobenzamide, 97%

Manufacturer:  Alfa Aesar™ H54550MD

 View more versions of this product

Catalog No. AAH54550MD



Specifications

Specifications

3-Methyl-4-nitrothiobenzamide
886364-22-3
MFCD06738330
3-methyl-4-nitrobenzene-1-carbothioamide
CC1=C(C=CC(=C1)C(=S)N)[N+](=O)[O-]
196.224
196.22
250mg
C8H8N2O2S
19566115
QNPFIXDGSZDAGC-UHFFFAOYSA-N
3-methyl-4-nitrobenzenecarbothioamide
55255791
97%

Chemical Identifiers

886364-22-3
196.224
QNPFIXDGSZDAGC-UHFFFAOYSA-N
55255791
CC1=C(C=CC(=C1)C(=S)N)[N+](=O)[O-]
C8H8N2O2S
MFCD06738330
3-methyl-4-nitrobenzene-1-carbothioamide
3-methyl-4-nitrobenzenecarbothioamide
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents